5-Brom-2-fluorpyridin

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS7142G100EA 0 CHF
ACTIAS7142G25 ACTIAS7142G100
5-Brom-2-fluorpyridin
5-Brom-2-fluorpyridin
Formel: C₅H₃BrFN
MW: 175,99 g/mol
Siedepunkt: 63 °C (15 mmHg)
MDL Number: MFCD01863742
CAS-Nummer: 766-11-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR