3-Brom-5-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS31249G1 AS31249G5
ACTIAS31249G1EA 0 CHF
ACTIAS31249G1 ACTIAS31249G5
3-Brom-5-methylpyridin
3-Brom-5-methylpyridin
Formel: C₆H₆BrN
MDL Number: MFCD01646141
CAS-Nummer: 3430-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR