3-Brom-5-methoxypyridin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 5-Bromo-3-methoxypyridine

AS50347G1 AS50347G5
ACTIAS50347G1EA 0 CHF
ACTIAS50347G1 ACTIAS50347G5
3-Brom-5-methoxypyridin
3-Bromo-5-methoxypyridine
Formel: C₆H₆BrNO
MW: 188,02 g/mol
MDL Number: MFCD00234169
CAS-Nummer: 50720-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR