3-Brom-5-fluorpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS4074G1 AS4074G5
ACTIAS4074G1EA 0 CHF
ACTIAS4074G1 ACTIAS4074G5
3-Brom-5-fluorpyridin
3-Bromo-5-fluoropyridine
Formel: C₅H₃BrFN
MW: 175.99 g/mol
MDL Number: MFCD04112555
CAS-Nummer: 407-20-5
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR