3-Brom-5-chlorpyridin
Lieferant: ACTIVATE SCIENTIFIC
AS51097G1
AS51097G5
ACTIAS51097G1EA
0
CHF
ACTIAS51097G1
ACTIAS51097G5
3-Brom-5-chlorpyridin
3-Bromo-5-chloropyridine
Formel:
C₅H₃BrClN MW: 192,44 g/mol |
MDL Number:
MFCD04114221 CAS-Nummer: 73583-39-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...