3-Brom-4-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 3-Bromo-4-picoline

AS50702G100 AS50702G5
ACTIAS50702G100EA 0 CHF
ACTIAS50702G100 ACTIAS50702G5
3-Brom-4-methylpyridin
3-Brom-4-methylpyridin
Formel: C₆H₆BrN
MW: 172,02 g/mol
Siedepunkt: 47…48 °C (0,6 mmHg)
Dichte: 1,549 g/cm³ (20 °C)
MDL Number: MFCD00082592
CAS-Nummer: 3430-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR