3-Brom-2-chlorbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS34128G1 AS34128G5
ACTIAS34128G1EA 0 CHF
ACTIAS34128G1 ACTIAS34128G5
3-Brom-2-chlorbenzoesäure
3-Bromo-2-chlorobenzoic acid
Formel: C₇H₄BrClO₂
MDL Number: MFCD01569399
CAS-Nummer: 56961-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR