3-Brom-2,5-dichlorpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Bromo-2,5-dichloro-pyridine

AS32590G1 AS32590G5
ACTIAS32590G1EA 0 CHF
ACTIAS32590G1 ACTIAS32590G5
3-Brom-2,5-dichlorpyridin
3-Bromo-2,5-dichloropyridine
Formel: C₅H₂BrCl₂N
MW: 226,89 g/mol
MDL Number: MFCD06659484
CAS-Nummer: 138006-41-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR