3-Amino-2,6-dichlorpyridin
Lieferant: ACTIVATE SCIENTIFIC
AS4069G1
AS4069G5
ACTIAS4069G1EA
0
CHF
ACTIAS4069G1
ACTIAS4069G5
3-Amino-2,6-dichlorpyridin
3-Amino-2,6-dichlorpyridin
Formel:
C₅H₄Cl₂N₂ Siedepunkt: 305 °C (1013 hPa) Melting Pt: 119 °C Dichte: 1.497 g/cm³ (20 °C) |
MDL Number:
MFCD00023417 CAS-Nummer: 62476-56-6 EINECS: 263-559-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...