3-Amino-2,5-dichlorpyridin

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS31574G1EA 0 CHF
ACTIAS31574G1 ACTIAS31574G5
3-Amino-2,5-dichlorpyridin
3-Amino-2,5-dichlorpyridin
Formel: C₅H₄Cl₂N₂
MW: 163,01 g/mol
Lager Temperatur: Kühlschrank
MDL Number: MFCD00204157
CAS-Nummer: 78607-32-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR