3,5-Difluorpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS50403G1 AS50403G100 AS50403G5
ACTIAS50403G1EA 0 CHF
ACTIAS50403G1 ACTIAS50403G100 ACTIAS50403G5
3,5-Difluorpyridin
3,5-Difluoropyridine
Formel: C₅H₃F₂N
MW: 115,08 g/mol
Siedepunkt: 92…93 °C (1013 hPa)
Dichte: 1.268 g/cm³ (20 °C)
MDL Number: MFCD03412222
CAS-Nummer: 71902-33-5
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR