3,4-Difluorphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3,4-Difluorobenzeneboronic acid, (3,4-Difluorophenyl)metaboric acid, (3,4-Difluorophenyl)boronic acid, Dihydroxy(3,4-difluorophenyl)borane

AS50256G1 AS50256G5
ACTIAS50256G1EA 0 CHF
ACTIAS50256G1 ACTIAS50256G5
3,4-Difluorphenylboronsäure
3,4-Difluorphenylboronsäure
Formel: C₆H₅BF₂O₂
MW: 157,91 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00807405
CAS-Nummer: 168267-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR