3,4-Dichlorphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
AS50214G100
AS50214G5
ACTIAS50214G100EA
0
CHF
ACTIAS50214G100
ACTIAS50214G5
3,4-Dichlorphenylboronsäure
3,4-Dichlorphenylboronsäure
Formel:
C₆H₅BCl₂O₂ MW: 190.82 g/mol |
MDL Number:
MFCD01074646 CAS-Nummer: 151169-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...