6-Chlor-o-toluylsäure

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS30960G1 AS30960G5
ACTIAS30960G1EA 0 CHF
ACTIAS30960G1 ACTIAS30960G5
6-Chlor-o-toluylsäure
6-Chlor-o-toluylsäure
Formel: C₈H₇ClO₂
MDL Number: MFCD00045799
CAS-Nummer: 21327-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR