2-Chlor-5-iodpyridin
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
AS31011G1
AS31011G5
ACTIAS31011G1EA
0
CHF
ACTIAS31011G1
ACTIAS31011G5
2-Chlor-5-iodpyridin
2-Chlor-5-iodpyridin
Formel:
C₅H₃ClIN MW: 239,44 g/mol |
MDL Number:
MFCD01863635 CAS-Nummer: 69045-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...