2-Chlor-5-hydroxypyridin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 2-Chloro-5-pyridinol, 6-Chloro-3-pyridinol

AS7203G100 AS7203G5
ACTIAS7203G100EA 0 CHF
ACTIAS7203G100 ACTIAS7203G5
2-Chlor-5-hydroxypyridin
2-Chlor-5-hydroxypyridin
Formel: C₅H₄ClNO
MW: 129,55 g/mol
MDL Number: MFCD00234050
CAS-Nummer: 41288-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR