6-Brom-o-toluylsäure
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
2-Bromo-6-methylbenzoic acid
AS33558G1
AS33558G10
AS33558G5
ACTIAS33558G1EA
0
CHF
ACTIAS33558G1
ACTIAS33558G10
ACTIAS33558G5
6-Brom-o-toluylsäure
6-Brom-o-toluylsäure
Formel:
C₈H₇BrO₂ |
MDL Number:
MFCD01310788 CAS-Nummer: 90259-31-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...