2-Brom-6-fluorpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS50220G1 AS50220G100 AS50220G5
ACTIAS50220G1EA 0 CHF
ACTIAS50220G1 ACTIAS50220G100 ACTIAS50220G5
2-Brom-6-fluorpyridin
2-Bromo-6-fluoropyridine
Formel: C₅H₃BrFN
MDL Number: MFCD03095258
CAS-Nummer: 144100-07-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR