6-Brom-m-toluylsäure
Lieferant: ACTIVATE SCIENTIFIC
AS31922G1
AS31922G5
ACTIAS31922G1EA
0
CHF
ACTIAS31922G1
ACTIAS31922G5
6-Brom-m-toluylsäure
6-Brom-m-toluylsäure
Formel:
C₈H₇BrO₂ MW: 215,05 g/mol |
MDL Number:
MFCD00079722 CAS-Nummer: 6967-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...