2-Brom-5-chlorpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS7189G100 AS7189G5
ACTIAS7189G100EA 0 CHF
ACTIAS7189G100 ACTIAS7189G5
2-Brom-5-chlorpyridin
2-Bromo-5-chloropyridine
Formel: C₅H₃BrClN
MDL Number: MFCD00234006
CAS-Nummer: 40473-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR