2-Amino-6-methyl-3-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Methyl-3-nitro-2-pyridylamine, 6-Methyl-3-nitropyridin-2-amine, 2-Amino-3-nitro-6-picoline, 2-Amino-3-nitro-6-picoline

AS31210G1 AS31210G5
ACTIAS31210G1EA 0 CHF
ACTIAS31210G1 ACTIAS31210G5
2-Amino-6-methyl-3-nitropyridin
2-Amino-6-methyl-3-nitropyridin
Formel: C₆H₇N₃O₂
MDL Number: MFCD00047443
CAS-Nummer: 21901-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR