6-Methoxyanthranilsäure

Lieferant: ACTIVATE SCIENTIFIC

AS20322G25 AS20322G5
ACTIAS20322G25EA 0 CHF
ACTIAS20322G25 ACTIAS20322G5
6-Methoxyanthranilsäure
6-Methoxyanthranilsäure
Formel: C₈H₉NO₃
Lager Temperatur: Raumtemperatur
MDL Number: MFCD01941328
CAS-Nummer: 53600-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR