2-Amino-5-fluorpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS7132G25 AS7132G5
ACTIAS7132G25EA 0 CHF
ACTIAS7132G25 ACTIAS7132G5
2-Amino-5-fluorpyridin
2-Amino-5-fluorpyridin
Formel: C₅H₅FN₂
MW: 112,11 g/mol
MDL Number: MFCD01861120
CAS-Nummer: 21717-96-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR