2,6-Dimethoxyphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
AS34929G1
AS34929G5
ACTIAS34929G1EA
0
CHF
ACTIAS34929G1
ACTIAS34929G5
2,6-Dimethoxyphenylboronsäure
2,6-Dimethoxyphenylboronsäure
Formel:
C₈H₁₁BO₄ MW: 181,98 g/mol |
MDL Number:
MFCD01318987 CAS-Nummer: 23112-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...