2,5-Dimethylindol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,5-Dimethyl-1H-indole

AS33684G1 AS33684G5
ACTIAS33684G1EA 0 CHF
ACTIAS33684G1 ACTIAS33684G5
2,5-Dimethylindol
2,5-Dimethylindol
Formel: C₁₀H₁₁N
MW: 145,2 g/mol
Siedepunkt: 282 °C (1013 hPa)
Melting Pt: 116 °C
Dichte: 1.08 g/cm³ (20 °C)
MDL Number: MFCD00005621
CAS-Nummer: 1196-79-8
EINECS: 214-816-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR