2,4-Dichlorphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
Dihydroxy(2,4-dichlorophenyl)borane, 2,4-Dichlorobenzeneboronic acid, (2,4-Dichlorophenyl)metaboric acid
AS50204G1
AS50204G100
AS50204G5
ACTIAS50204G1EA
0
CHF
ACTIAS50204G1
ACTIAS50204G100
ACTIAS50204G5
2,4-Dichlorphenylboronsäure
2,4-Dichlorphenylboronsäure
Formel:
C₆H₅BCl₂O₂ MW: 190,82 g/mol |
MDL Number:
MFCD00013930 CAS-Nummer: 68716-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...