2,4,6-Trifluorbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS33281G1 AS33281G10 AS33281G5
ACTIAS33281G1EA 0 CHF
ACTIAS33281G1 ACTIAS33281G10 ACTIAS33281G5
2,4,6-Trifluorbenzaldehyd
2,4,6-Trifluorobenzaldehyde
Formel: C₇H₃F₃O
MW: 160,1 g/mol
MDL Number: MFCD00061197
CAS-Nummer: 58551-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR