2,3,6-Trifluorphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS51593G1 AS51593G5
ACTIAS51593G1EA 0 CHF
ACTIAS51593G1 ACTIAS51593G5
2,3,6-Trifluorphenylboronsäure
2,3,6-Trifluorphenylboronsäure
Formel: C₆H₄BF₃O₂
MDL Number: MFCD01863167
CAS-Nummer: 247564-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR