3-Fluor-4-nitroanisol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoro-4-methoxy-1-nitrobenzene

AS31294G1 AS31294G5
ACTIAS31294G1EA 0 CHF
ACTIAS31294G1 ACTIAS31294G5
3-Fluor-4-nitroanisol
3-Fluor-4-nitroanisol
Formel: C₇H₆FNO₃
MW: 171,13 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD04115632
CAS-Nummer: 446-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR