3-Fluor-p-kresol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Fluoro-4-methylphenol, 1-Fluoro-3-hydroxy-6-methylbenzene

AS32381G1 AS32381G5
ACTIAS32381G1EA 0 CHF
ACTIAS32381G1 ACTIAS32381G5
3-Fluor-p-kresol
3-Fluor-p-kresol
Formel: C₇H₇FO
MDL Number: MFCD00143164
CAS-Nummer: 452-78-8
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR