3,4,5-Trifluorbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS22441G100 AS22441G25
ACTIAS22441G100EA 0 CHF
ACTIAS22441G100 ACTIAS22441G25
3,4,5-Trifluorbenzonitril
3,4,5-Trifluorbenzonitril
Formel: C₇H₂F₃N
Siedepunkt: 72…73 °C (10 mmHg)
Melting Pt: 45…47 °C
MDL Number: MFCD00074948
CAS-Nummer: 134227-45-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR