3-Chlor-2-nitrotoluol
Lieferant: ACTIVATE SCIENTIFIC
AS22250G25
AS22250G5
ACTIAS22250G25EA
156
CHF
ACTIAS22250G25
ACTIAS22250G5
3-Chlor-2-nitrotoluol
3-Chlor-2-nitrotoluol
Formel:
C₇H₆ClNO₂ MW: 171,58 g/mol |
MDL Number:
MFCD04117977 CAS-Nummer: 5367-26-0 EINECS: 226-354-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...