1-Chlor-3-fluor-2-nitrobenzol
Lieferant: ACTIVATE SCIENTIFIC
AS31963G1
AS31963G5
ACTIAS31963G1EA
0
CHF
ACTIAS31963G1
ACTIAS31963G5
1-Chlor-3-fluor-2-nitrobenzol
Formel:
C₆H₃ClFNO₂ |
MDL Number:
MFCD06658264 CAS-Nummer: 64182-61-2 EINECS: 264-723-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...