2-Chlor-6-fluoranisol

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS40217G1 AS40217G5
ACTIAS40217G1EA 0 CHF
ACTIAS40217G1 ACTIAS40217G5
2-Chlor-6-fluoranisol
2-Chlor-6-fluoranisol
Formel: C₇H₆ClFO
MW: 160,58 g/mol
MDL Number: MFCD01631573
CAS-Nummer: 53145-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR