5-Brom-α,α,α,2-tetrafluortoluol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Bromo-2-fluorobenzotrifluoride, 1-Bromo-4-fluoro-3-(trifluoromethyl)benzene

AS21904G100 AS21904G25
ACTIAS21904G100EA 0 CHF
ACTIAS21904G100 ACTIAS21904G25
5-Brom-α,α,α,2-tetrafluortoluol
5-Brom-α,α,α,2-tetrafluortoluol
Formel: C₇H₃BrF₄
MW: 243 g/mol
MDL Number: MFCD00042493
CAS-Nummer: 393-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR