3-Brom-p-kresol
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
3-Bromo-4-methylphenol, 1-Bromo-3-hydroxy-6-methylbenzene
AS32427G1
AS32427G5
ACTIAS32427G1EA
0
CHF
ACTIAS32427G1
ACTIAS32427G5
3-Brom-p-kresol
3-Brom-p-kresol
Formel:
C₇H₇BrO MW: 187,04 g/mol |
MDL Number:
MFCD08273793 CAS-Nummer: 60710-39-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...