5-Brom-o-kresol
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
AS30682G1
AS30682G5
ACTIAS30682G1EA
0
CHF
ACTIAS30682G1
ACTIAS30682G5
5-Brom-o-kresol
5-Brom-o-kresol
Formel:
C₇H₇BrO |
MDL Number:
MFCD08061906 CAS-Nummer: 36138-76-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...