4-Brom-2,6-dichlorphenol

Lieferant: ACTIVATE SCIENTIFIC

AS51397G1 AS51397G5
ACTIAS51397G1EA 0 CHF
ACTIAS51397G1 ACTIAS51397G5
4-Brom-2,6-dichlorphenol
4-Brom-2,6-dichlorphenol
Formel: C₆H₃BrCl₂O
MW: 241,9 g/mol
MDL Number: MFCD00019996
CAS-Nummer: 3217-15-0
EINECS: 221-740-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR