5-Brom-o-toluidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Bromo-2-methylaniline, 5-Bromo-o-toluidine, 2-Amino-4-bromotoluene

AS33805G10 AS33805G100
ACTIAS33805G10EA 0 CHF
ACTIAS33805G10 ACTIAS33805G100
5-Brom-o-toluidin
5-Brom-o-toluidin
Formel: C₇H₈BrN
Siedepunkt: 260 °C (1013 hPa)
Melting Pt: 33 °C
Dichte: 1.49 g/cm³ (25 °C)
MDL Number: MFCD00800678
CAS-Nummer: 39478-78-9
EINECS: 254-467-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR