5-Fluor-o-toluidin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Amino-4-fluorotoluene, 5-Fluoro-2-methylaniline
AS33792G10
AS33792G100
ACTIAS33792G10EA
0
CHF
ACTIAS33792G10
ACTIAS33792G100
5-Fluor-o-toluidin
5-Fluor-o-toluidin
Formel:
C₇H₈FN MW: 125,15 g/mol Siedepunkt: 203 °C (1013 hPa) Melting Pt: 36…38 °C Dichte: 1.13 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00007764 CAS-Nummer: 367-29-3 EINECS: 206-689-6 UN: 2811 ADR: 6.1,III |
Learn more
![](/m-ch.vwr.com/de_CH/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...