3-Brom-o-toluidin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 2-Amino-6-bromotoluene, 3-Bromo-2-methylaniline

AS21032G100 AS21032G25
ACTIAS21032G100EA 0 CHF
ACTIAS21032G100 ACTIAS21032G25
3-Brom-o-toluidin
3-Brom-o-toluidin
Formel: C₇H₈BrN
Siedepunkt: 253…255 °C (1013 hPa)
Dichte: 1.51 g/cm³ (20 °C)
MDL Number: MFCD00051579
CAS-Nummer: 55289-36-6
EINECS: 259-568-5
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR