2-Amino-3-nitrophenol
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
2-Hydroxy-6-nitroaniline
AS20215G100
AS20215G25
ACTIAS20215G100EA
0
CHF
ACTIAS20215G100
ACTIAS20215G25
2-Amino-3-nitrophenol
2-Amino-3-nitrophenol
Formel:
C₆H₆N₂O₃ MW: 154.13 g/mol |
MDL Number:
MFCD00010875 CAS-Nummer: 603-85-0 EINECS: 210-060-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...