2-Brom-5-fluoranilin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS33673G10 AS33673G100
ACTIAS33673G10EA 0 CHF
ACTIAS33673G10 ACTIAS33673G100
2-Brom-5-fluoranilin
2-Brom-5-fluoranilin
Formel: C₆H₅BrFN
MW: 190.01 g/mol
MDL Number: MFCD00070750
CAS-Nummer: 1003-99-2
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR