1,8-Dihydroxynaphthalin

Lieferant: ACTIVATE SCIENTIFIC

AS34822G1 AS34822G5
ACTIAS34822G1EA 0 CHF
ACTIAS34822G1 ACTIAS34822G5
1,8-Dihydroxynaphthalin
1,8-Dihydroxynaphthalin
Formel: C₁₀H₈O₂
MW: 160.17 g/mol
MDL Number: MFCD00042701
CAS-Nummer: 569-42-6
EINECS: 209-316-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR