2',5'-Difluoracetophenon

Lieferant: ACTIVATE SCIENTIFIC

AS34447G10 AS34447G100
ACTIAS34447G10EA 0 CHF
ACTIAS34447G10 ACTIAS34447G100
2',5'-Difluoracetophenon
2',5'-Difluoracetophenon
Formel: C₈H₆F₂O
Siedepunkt: 70…80 °C (15 mmHg)
Dichte: 1,2099 g/cm³ (22 °C)
MDL Number: MFCD00009898
CAS-Nummer: 1979-36-8
EINECS: 217-837-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR