2,6-Difluorbenzylalkohol
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
(2,6-Difluorophenyl)methanol
AS22372G100
AS22372G25
ACTIAS22372G100EA
105
CHF
ACTIAS22372G100
ACTIAS22372G25
2,6-Difluorbenzylalkohol
2,6-Difluorbenzylalkohol
Formel:
C₇H₆F₂O MW: 144,12 g/mol Siedepunkt: 88 °C (14 mmHg) Dichte: 1,3 g/cm³ (20 °C) |
MDL Number:
MFCD00004603 CAS-Nummer: 19064-18-7 EINECS: 242-792-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...