L(+)-Leucin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
(S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine
AS33094G1
AS33094G5
ACTIAS33094G1EA
0
CHF
ACTIAS33094G1
ACTIAS33094G5
L(+)-Leucin
L(+)-Leucin
Formel:
C₆H₁₃NO₂ MW: 131,17 g/mol Melting Pt: 300 °C Dichte: 1.165 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00002617 CAS-Nummer: 61-90-5 EINECS: 200-522-0 Merck Index: 14,05451 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...