(2-Chlor-6-fluorphenyl)essigsäure

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 2-Chloro-6-fluorophenylacetic acid

AS35283G1 AS35283G10 AS35283G5
ACTIAS35283G1EA 0 CHF
ACTIAS35283G1 ACTIAS35283G10 ACTIAS35283G5
(2-Chlor-6-fluorphenyl)essigsäure
(2-Chlor-6-fluorphenyl)essigsäure
Formel: (C₆H₃ClF)CH₂COOH
MW: 188,59 g/mol
Siedepunkt: 289 °C (1013 hPa)
Melting Pt: 120…123 °C
Dichte: 1 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004319
CAS-Nummer: 37777-76-7
EINECS: 253-661-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR