(2,4,5-Trifluorphenyl)essigsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,4,5-Trifluorophenylacetic acid

AS20228G100 AS20228G25
ACTIAS20228G100EA 56 CHF
ACTIAS20228G100 ACTIAS20228G25
(2,4,5-Trifluorphenyl)essigsäure
(2,4,5-Trifluorphenyl)essigsäure
Formel: (C₆H₂F₃)CH₂COOH
MW: 190,12 g/mol
MDL Number: MFCD00082479
CAS-Nummer: 209995-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR