5-Methylpyrimidin

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS31987G1 AS31987G5
ACTIAS31987G1EA 0 CHF
ACTIAS31987G1 ACTIAS31987G5
5-Methylpyrimidin
5-Methylpyrimidine
Formel: C₅H₆N₂
MW: 94,12 g/mol
Siedepunkt: 153 °C (1013 hPa)
Melting Pt: 28 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00047386
CAS-Nummer: 2036-41-1
EINECS: 218-006-9
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR